missing translation for 'onlineSavingsMsg'
Learn More

Brady™ B30 Series Metallized Polyester Label Material

Catalog No. 19100027
Change view
Click to view available options
Adhesive Type:
Acrylic, Permanent
Permanent
Finish:
Gloss
Matte
Thickness (English):
0.004 in.
0.0044 in.
0.0053 in.
Temperature Range (English):
-40° to +248°F
-40° to +194°F
-40° to +248°F
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Adhesive Type Finish Thickness (English) Temperature Range (English)
19100027 Acrylic, Permanent Matte 0.0044 in. -40° to +248°F
19100029 Acrylic, Permanent Gloss 0.004 in. -40° to +194°F
19100040 Permanent Matte 0.0053 in. -40° to +248°F
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19100027 Supplier Brady™ Supplier No. B30C4000428

Designed to withstand numerous solvents and variable temperatures

  • Metallized polyester material
  • Matte or glossy finish
  • Permanent acrylic or rubber-based adhesive
  • For applications like labelling rating and serial plates

Specifications

Adhesive Type Acrylic, Permanent
Finish Matte
Material Metallized Polyester
Temperature Range (English) -40° to +248°F
Thickness (English) 0.0044 in.