missing translation for 'onlineSavingsMsg'
Learn More

b-Nicotinamide Adenine Dinucleotide, 95%, Spectrum™ Chemical

Catalog No. 18601039
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601039 1 kg Amber Glass Bottle
18601035 1 g Amber Glass Bottle
18601036 5 g Amber Glass Bottle
18601037 25 g Amber Glass Bottle
18601038 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18601039 Supplier Spectrum Chemical Mfg Cor Supplier No. NI1021KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

NI102, 53-84-9, C21H27N7O14P2·3H2O

Spectrum™ Chemical b-Nicotinamide Adenine Dinucleotide

Chemical Identifiers

CAS 53-84-9
Molecular Formula C21H27N7O14P2
Molecular Weight (g/mol) 663.43
MDL Number MFCD00150377
InChI Key BAWFJGJZGIEFAR-WIWLTUSXNA-N
IUPAC Name 1-[(2R,3R,4S,5R)-5-{[({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]methyl}-3,4-dihydroxyoxolan-2-yl]-3-carbamoyl-1λ5-pyridin-1-ylium
SMILES NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 1 kg
Percent Purity 95%
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material b-Nicotinamide Adenine Dinucleotide