Learn More
Gibco™ B-27™ Supplement (50X), serum free
Description
This original and genuine B-27 Supplement is a cGMP-manufactured, serum-free solution designed to ensure the consistent, reliable, and high-quality growth of embryonic, post-natal, and adult hippocampal neurons, as well as other CNS neurons. With a proven track record, B-27 Supplement supports both low- and high-density cultures, ensuring viability in serum-free media. Each batch undergoes rigorous performance testing to confirm its ability to successfully culture hippocampal neurons, providing you with the confidence and dependability you need for your research.
- Dependable consistency—every lot is performance-tested for reliable results
- High quality—cGMP manufacturing provides the highest standards of quality
- Versatile usage—ideal for use with serum-free media such as Neurobasal Medium or Neurobasal-A Medium, without the need for an astrocyte feeder layer. Supports the growth and viability of neurons, as well as many other cell types differentiated from PSCs.
- Proven performance—a proprietary formulation; the first and most cited neuronal cell culture supplement, trusted by researchers worldwide
Choose B-27 Supplement for the authentic solution that sets the standard for neuronal cell culture.
Order Info
Dry Ice
Spécifications
Spécifications
| Content And Storage | Store in freezer (-5 to -20°C) and protect from light. |
| Cell Type | Neural Cells |
| Classification | Serum-free |
| Concentration | 50 X |
| Culture Type | Mammalian Cell Culture |
| Form | Liquid |
| Product Type | Cell Culture Supplement |
| Serum Level | Serum-Free |
| Sterility | Sterile |
| pH | 6 to 8 |
| Afficher plus de résultats |
For Research Use Only. Not for use in diagnostic procedures.