missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azure Biosystems AzureRed Fluorescent Protein Stain
Description
- Quantitative fluorescent total protein stain for gels and Western blots
- For use with Western blot samples of biological fluids with very low protein concentration, 0.1 to 1μg lysate/lane. Choose AzureRed Fluorescent Protein Stain for sensitive detection of total protein on 1D or 2D gels. AzureRed is as sensitive as silver stain and is compatible with downstream Western blotting, mass spectrometry, and Edman sequencing. Because the free dye is non-fluorescent, AzureRed provides very low background and higher signal to noise than other fluorescent stains. AzureRed-stained gels can be imaged with UV or blue light excitation.
- Allows you to detect less than 1 ng of protein per band or spot. Non-toxic and biodegradable, for safe and simple disposal. Provided as a 200x concentrated solution.
- Each kit contains:
- 5 mL AzureRed Fluorescent Protein Stain
- AzureRed Powder A, for fix solution preparation
- AzureRed Powder B, for stain buffer preparation
Specifications
Specifications
| Color | Clear |
| Concentration | 200X |
| Detection Method | Fluorescense |
| Format | Bottle |
| Formulation | Liquid |
| Storage Requirements | Room Temperature |
| Product Type | Total Protein Stain for Nitrocellulose |