missing translation for 'onlineSavingsMsg'
Learn More

Aztreonam, MilliporeSigma™ Supelco™

Catalog No. 111015538
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015538 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015538 Supplier Merck Emd Millipore Supplier No. A684850MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Aztreonam is a synthetic monocyclic β-lactum, which can be isolated from Chromobacterium violaceum. It may find clinical applications as a potent antimicrobial agent, due to its bactericidal action.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 78110-38-0
Physical Form Powder/Crystals
Linear Formula C13H17N5O8S2
MDL Number MFCD00072145
Synonym [2 S-[2 alpha,3 beta(Z)]]-2-[[[1-(2-Amino-4-thiazolyl)-2-[(2-methyl-4-oxo-1-sulfo-3-azetidinyl)amino]-2-oxoethylidene]amino]oxy]-2-methylpropanoic acid
Type Analytical Standard
Quantity 50 mg
Formula Weight 435.43
Grade Analytical Standard