missing translation for 'onlineSavingsMsg'
Learn More

Azoic Diazo Component 22 98.0+%, TCI America™

Catalog No. P0203500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0203500G 500 g
P020325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0203500G Supplier TCI America Supplier No. P0203500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101-54-2
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.242
MDL Number MFCD00007850
InChI Key ATGUVEKSASEFFO-UHFFFAOYSA-N
Synonym 4-aminodiphenylamine, n-phenyl-p-phenylenediamine, n1-phenylbenzene-1,4-diamine, p-anilinoaniline, azosalt r, p-semidine, p-aminodiphenylamine, luxan black r, semidine, n-phenyl-1,4-phenylenediamine
PubChem CID 7564
ChEBI CHEBI:59038
IUPAC Name 4-N-phenylbenzene-1,4-diamine
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)N

Specifications

Melting Point 74°C
Color Black-Gray
Boiling Point 354°C
Quantity 500 g
Formula Weight 184.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Azoic Diazo Component 22
EINECSNumber (3)-0134&(3)-2720&(5)-2313
RTECSNumber ST3150000
TSCA Yes