Learn More
Abcam Inc Azithromycin dihydrate, Macrolide antibiotic, 50MG
Numéro de catalogue. 502706939
Encompass
MW 749 Da. Macrolide antibiotic. Bacterial 50S ribosomal inhibitor. Inhibits translation, cell viability and growth rate of bacterial cells (IC = 0.4 g/ml). Orally active against gram-negative and gram-positive bacteria.
The product is subject to the following: Abcam Restricted Use Statement
Numéro de catalogue. 502706939 Fournisseur Abcam Inc Code fournisseur AB14106450MG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
MW 749 Da. Macrolide antibiotic. Bacterial 50S ribosomal inhibitor. Inhibits translation, cell viability and growth rate of bacterial cells (IC = 0.4 g/ml). Orally active against gram-negative and gram-positive bacteria.
The product is subject to the following: Abcam Restricted Use Statement