missing translation for 'onlineSavingsMsg'
Learn More

Azido-PEG4-NHS Ester, TCI America™

Catalog No. A238825MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A238825MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A238825MG Supplier TCI America Supplier No. A238825MG

Chemical Identifiers

CAS 944251-24-5
Molecular Formula C15H24N4O8
Molecular Weight (g/mol) 388.377
InChI Key OIGKWPIMJCPGGD-UHFFFAOYSA-N
Synonym NHS-PEG4-Azide, N-Succinimidyl 15-Azido-4,7,10,13-tetraoxapentadecanoate, 15-Azido-4,7,10,13-tetraoxapentadecanoic Acid N-Succinimidyl Ester
PubChem CID 51340931
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 3-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethoxy]propanoate
SMILES C1CC(=O)N(C1=O)OC(=O)CCOCCOCCOCCOCCN=[N+]=[N-]

Specifications

Color Colorless
Quantity 25 mg
Formula Weight 388.38
Physical Form Liquid
Chemical Name or Material Azido-PEG4-NHS Ester
TSCA No
Recommended Storage Freezer