missing translation for 'onlineSavingsMsg'
Learn More

Azido-PEG3-biotin conjugate

Catalog No. AAJ64996MB
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64996MB 25 mg
AAJ64996MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64996MB Supplier Thermo Scientific Chemicals Supplier No. J64996MB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An azide-biotin conjugate with PEG spacer to improve solubility in aqueous media | CAS: 875770-34-6 | C18H32N6O5S | 444.551 g/mol

Biotinylation reagent for labeling alkyne containing molecules or biomolecules using either copper-catalyzed 1,3 dipolar cycloaddition click chemistry or copper-free click chemistry with cyclooctyne derivatives. The azide group reacts with alkynes to form a stable triazole linkage facilitating the introduction of biotin into your alkyne modified system of interest. This reaction is bioorthogonal or in other words compatible with biological systems in that it?s components do not react with the biological environment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biotinylation reagent for labeling alkyne containing molecules or biomolecules using either copper-catalyzed 1,3 dipolar cycloaddition click chemistry or copper-free click chemistry with cyclooctyne derivatives. The azide group reacts with alkynes to form a stable triazole linkage facilitating the introduction of biotin into your alkyne modified system of interest. This reaction is bioorthogonal or in other words compatible with biological systems in that it′s components do not react with the biological environment.

Solubility
Solubile in DMSO, DMF and water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 875770-34-6
Molecular Formula C18H32N6O5S
Molecular Weight (g/mol) 444.551
MDL Number MFCD20134145
InChI Key ZWFOOMQCYIGZBE-ZOBUZTSGSA-N
Synonym biotin-peg3-azide, n-2-2-2-2-azidoethoxy ethoxy ethoxy ethyl biotinamide, n-2-2-2-2-azidoethoxy ethoxy ethoxy ethyl-5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanamide, biotin-peg4-n3, azide-peg3-biotin conjugate, azido-peg3-biotin conjugate, 11-r-+-biotinylamino-1-azido-3,6,9-trioxaundecane, 3abeta,6abeta-n-2-2-2-2-azidoethoxy ethoxy ethoxy ethyl-2-oxohexahydro-1h-thieno 3,4-d imidazole-4alpha-pentanamide
PubChem CID 60146223
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethyl]pentanamide
SMILES C1C2C(C(S1)CCCCC(=O)NCCOCCOCCOCCN=[N+]=[N-])NC(=O)N2

Specifications

Color White to Gray
Quantity 25 mg
Solubility Information Solubile in DMSO,DMF and water.
Formula Weight 444.55
Physical Form Solid
Chemical Name or Material Azido-PEG3-biotin conjugate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only