missing translation for 'onlineSavingsMsg'
Learn More

Azetidine hydrochloride, 97%

Catalog No. AAH5993506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5993506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5993506 Supplier Thermo Scientific Chemicals Supplier No. H5993506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 36520-39-5 | C3H8ClN | 93.554 g/mol

Azetidine hydrochloride is a building block for azetidine synthesis and a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azetidine hydrochloride is a building block for azetidine synthesis and a pharmaceutical intermediate.

Solubility
Soluble in water (103.8°C at 760 mmHg).

Notes
Hygroscopic. Reacts with strong oxidizing agents.

Chemical Identifiers

CAS 36520-39-5
Molecular Formula C3H8ClN
Molecular Weight (g/mol) 93.554
MDL Number MFCD00191762
InChI Key HGQULGDOROIPJN-UHFFFAOYSA-N
Synonym azetidine hydrochloride, azetidine hcl, trimethyleneimine hydrochloride, azetidine, hydrochloride, azetidinehydrochloride, azetidine-hcl, azetidine hcl salt, azetidine-hydrochloride, azetidine; hydrochloride, acmc-1aiho
PubChem CID 12308726
IUPAC Name azetidine;hydrochloride
SMILES C1CNC1.Cl

Specifications

Melting Point >300°C
Flash Point 152°C (306°F)
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water (103.8°C at 760mmHg).
Formula Weight 93.56
Percent Purity 97%
Chemical Name or Material Azetidine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only