missing translation for 'onlineSavingsMsg'
Learn More

Azetidine-3-carboxylic Acid 96.0+%, TCI America™

Catalog No. A1646100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1646100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1646100MG Supplier TCI America Supplier No. A1646100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 36476-78-5
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00191763
InChI Key GFZWHAAOIVMHOI-UHFFFAOYSA-N
Synonym 3-azetidinecarboxylic acid, 3-azetanecarboxylic acid, 3-carboxyazetidine, caswell no. 063c, h-aze 3-oh, azetane-3-carboxylic acid, epa pesticide chemical code 128830, 3-azetidine carboxylic acid, l-azetidine-3-carboxylic acid, azetidine-3-carboxylicacid
PubChem CID 93192
IUPAC Name azetidine-3-carboxylic acid
SMILES OC(=O)C1CNC1

Specifications

Melting Point 275°C
Color White-Yellow
Quantity 100 mg
Formula Weight 101.11
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Azetidine-3-carboxylic Acid
RTECSNumber CM4310600
TSCA No