missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azer Scientific Blood Bank Saline
For use in immunohematologic testing
$66.48 - $79.15
Specifications
| Certifications/Compliance | NCCLS |
|---|---|
| For Use With (Application) | For use in immunohematologic testing |
| Includes | Spigot |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Type | pH | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Type | pH | Price | Quantity & Availability | |||
|
22026400
|
Azer Scientific
ES124410L |
10,000 mL | 0.85% Non-buffered | 6.5 to 7 |
Each for $66.48
|
|
|||
|
22026401
|
Azer Scientific
ES124420L |
20,000 mL | 0.85% Non-buffered | 6.5 to 7 |
Each for $79.15
|
|
|||
|
22026403
|
Azer Scientific
ES124320L |
20,000 mL | 0.85% Buffered | 7 to 7.2 |
Each for $79.15
|
|
|||
|
22026406
|
Azer Scientific
ES124310L |
10,000 mL | 0.85% Buffered | 6.5 to 7 |
Each for $66.48
|
|
|||
Description
0.85% saline solutions meet NCCLS specifications for standard isotonic sodium chloride solution for use in immunohematologic testing. Standard and buffered solutions are available. Buffered saline is ideal for enhancing weak agglutination reactions in blood bank procedures. pH ranges are controlled.
pH Ranges
- Non-Buffered Saline: 6.5 - 7.0
- Buffered Saline: 7.0 - 7.2
For use in immunohematologic testing
Certifications
Meets NCCLS specifications for standard isotonic NaCl solution
Specifications
| NCCLS | |
| For use in immunohematologic testing | |
| Spigot | |
| Liquid |