missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azer Scientific Blood Bank Saline
Description
0.85% saline solutions meet NCCLS specifications for standard isotonic sodium chloride solution for use in immunohematologic testing. Standard and buffered solutions are available. Buffered saline is ideal for enhancing weak agglutination reactions in blood bank procedures. pH ranges are controlled.
pH Ranges
- Non-Buffered Saline: 6.5 - 7.0
- Buffered Saline: 7.0 - 7.2
For use in immunohematologic testing
Certifications
Meets NCCLS specifications for standard isotonic NaCl solution
Specifications
Specifications
| Certifications/Compliance | NCCLS |
| Type | 0.85% Non-buffered |
| For Use With (Application) | For use in immunohematologic testing |
| Includes | Spigot |
| pH | 6.5 to 7 |
| Quantity | 20,000 mL |
| Form | Liquid |