missing translation for 'onlineSavingsMsg'
Learn More

Azer Scientific Blood Bank Saline

Catalog No. 22026400
Change view
Click to view available options
Quantity:
10,000 mL
20,000 mL
Type:
0.85% Buffered
0.85% Non-buffered
pH:
6.5 to 7
7 to 7.2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type pH
22026400 10,000 mL 0.85% Non-buffered 6.5 to 7
22026401 20,000 mL 0.85% Non-buffered 6.5 to 7
22026403 20,000 mL 0.85% Buffered 7 to 7.2
22026406 10,000 mL 0.85% Buffered 6.5 to 7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22026400 Supplier Azer Scientific Supplier No. ES124410L

For use in immunohematologic testing

0.85% saline solutions meet NCCLS specifications for standard isotonic sodium chloride solution for use in immunohematologic testing. Standard and buffered solutions are available. Buffered saline is ideal for enhancing weak agglutination reactions in blood bank procedures. pH ranges are controlled.

pH Ranges

  • Non-Buffered Saline: 6.5 - 7.0
  • Buffered Saline: 7.0 - 7.2

For use in immunohematologic testing

Certifications

Meets NCCLS specifications for standard isotonic NaCl solution

Specifications

Certifications/Compliance NCCLS
Type 0.85% Non-buffered
For Use With (Application) For use in immunohematologic testing
Includes Spigot
pH 6.5 to 7
Quantity 10,000 mL
Form Liquid