missing translation for 'onlineSavingsMsg'
Learn More

Azelastine Hydrochloride 98.0+%, TCI America™

Catalog No. A23401G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23401G 1 g
A2340100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23401G Supplier TCI America Supplier No. A23401G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79307-93-0
Molecular Formula C22H25Cl2N3O
Molecular Weight (g/mol) 418.36
MDL Number MFCD00242783
InChI Key YEJAJYAHJQIWNU-UHFFFAOYNA-N
Synonym 4-(4-Chlorobenzyl)-2-(1-methylazepan-4-yl)-1-phthalazinone Hydrochloride
PubChem CID 50986914
ChEBI CHEBI:2951
IUPAC Name hydrogen 4-[(4-chlorophenyl)methyl]-2-(1-methylazepan-4-yl)-1,2-dihydrophthalazin-1-one chloride
SMILES [H+].[Cl-].CN1CCCC(CC1)N1N=C(CC2=CC=C(Cl)C=C2)C2=CC=CC=C2C1=O

Specifications

Melting Point 226°C
Color White
Quantity 1 g
Formula Weight 418.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Azelastine Hydrochloride
RTECSNumber TH9203900
TSCA No
Recommended Storage Freezer