missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azelaic acid, 98%
Description
Useful in lubrication and plasticizers. Azelaic acid is used to treat acne.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | ∼102°C |
| Boiling Point | 286°C (100mmHg) |
| Density | 1.225 g/mL |
| Quantity | 5 x 100 g |
| Flash Point | 215°C (419°F) |
| Physical Form | Crystalline |
| Beilstein | 1101094 |
| Merck Index | 14,905 |
| Solubility Information | Soluble in water |
| Formula Weight | 188.22 |
| Show More |
Safety and Handling
| EINECSNumber | 204-669-1 |
| RTECSNumber | CM1980000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only