missing translation for 'onlineSavingsMsg'
Learn More

Azathioprine

Catalog No. AAJ6231409
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6231409 10 g
AAJ6231403 1 g
AAJ6231406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6231409 Supplier Thermo Scientific Chemicals Supplier No. J6231409

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 446-86-6 | C9H7N7O2S | 277.262 g/mol

Azathioprine is an immunosuppressive purine antimetabolite. The mechanism of action of azathioprine is as a nucleic acid synthesis inhibitor. It has shown promise in treatment of alopecia areata (autoimmune hair loss), with no difference in effectiveness between genders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in Dichloromethane and dimethyl sulfoxide.

Notes
Store at -20°C. Incompatible with oxidizing agents. Protect from heat.

Chemical Identifiers

CAS 446-86-6
Molecular Formula C9H7N7O2S
Molecular Weight (g/mol) 277.262
MDL Number MFCD00069203
InChI Key LMEKQMALGUDUQG-UHFFFAOYSA-N
Synonym azathioprine, azothioprine, imuran, azathioprin, azatioprin, azamun, azanin, imurel, ccucol, imurek
PubChem CID 2265
ChEBI CHEBI:2948
IUPAC Name 6-(3-methyl-5-nitroimidazol-4-yl)sulfanyl-7H-purine
SMILES CN1C=NC(=C1SC2=NC=NC3=C2NC=N3)[N+](=O)[O-]

Specifications

Color Yellow
Quantity 10 g
Merck Index 14,902
Solubility Information Soluble in Dichloromethane and dimethyl sulfoxide.
Formula Weight 277.26
Physical Form Crystalline Powder
Chemical Name or Material Azathioprine
Hazard Category H302-H315-H319-H335-H350
Hazard Statement GHS H Statement
H350-H302-H315-H319-H335
May cause cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
EINECSNumber 207-175-4
RTECSNumber UO8925000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only