missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AZ 3146

Catalog No. 399410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
399410 10 mg
399450 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 399410 Supplier Tocris Bioscience™ Supplier No. 3994/10

Potent and selective Mps1 kinase inhibitor

Potent and selective monopolar spindle 1 (Mps1) kinase inhibitor (IC50 = 35 nM). Displays selectivity over 46 other kinases including Cdk1 and aurora kinase B. Interferes with chromosome alignment and overrides spindle assembly checkpoint. Inhibits the recruitment of Mad1, Mad2 and centromere protein E (CENP-E) to kinetochores.

Chemical Identifiers

CAS 1124329-14-1
Molecular Formula C24H32N6O3

Specifications

Quantity 10 mg
Formula Weight 452.55
Purity >99%
Molecular Weight (g/mol) 452.559
Chemical Name or Material AZ 3146
Recommended Storage Store at +4°C