missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ AxyPrep™ Mag Viral NA

The Axygen™ AxyPrep MAG Viral Nucleic Acid Purification Kit utilizes a unique paramagnetic beads-based system for the purification of viral nucleic acid from samples such as plasma, serum, ascites, cell culture supernatant, cerebrospinal fluid, and urine.

Specifications

For Use With (Application) Viral RNA/DNA Extraction
View More Specs

Products 2
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
14222382
View Documents Promotion Details
Axygen™
MAGVNAM
384 Preps N/A
Only null left
 
14222381
View Documents Promotion Details
Axygen™
MAGVNAS
96 Preps N/A
Only null left
 
Description

Description

Suitable for the following downstream applications:

  • PCR including RT-PCR and RT-qPCR
  • Sanger and next generation sequencing
  • Genotyping and SNP detection
  • Gene expression

Features:

  • Simple extraction process
  • Compatible with genomics downstream applications
  • High quality, high yield nucleic acid purification
  • Can be automated or used manually
Specifications

Specifications

Viral RNA/DNA Extraction
Videos
SDS
Documents

Documents