missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ AxyPrep™ Mag Viral NA

Catalog No. 14222382
Change view
Click to view available options
No. of Reactions:
384 Preps
96 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
14222382 384 Preps
14222381 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14222382 Supplier Axygen™ Supplier No. MAGVNAM
Only null left

The Axygen™ AxyPrep MAG Viral Nucleic Acid Purification Kit utilizes a unique paramagnetic beads-based system for the purification of viral nucleic acid from samples such as plasma, serum, ascites, cell culture supernatant, cerebrospinal fluid, and urine.

Suitable for the following downstream applications:

  • PCR including RT-PCR and RT-qPCR
  • Sanger and next generation sequencing
  • Genotyping and SNP detection
  • Gene expression

Features:

  • Simple extraction process
  • Compatible with genomics downstream applications
  • High quality, high yield nucleic acid purification
  • Can be automated or used manually

Specifications

For Use With (Application) Viral RNA/DNA Extraction
No. of Reactions 384 Preps