missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Oasis™ Robotic Tips

For use with Oasis automated liquid handling systems

$419.09 - $2248.04

Specifications

Sterility Non-sterile
Color Clear
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
View More Specs

Products 5
Catalog Number Mfr. No. Volume (Metric) Rack Color Filtered Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Rack Color Filtered Price Quantity  
14-222-220
View Documents
Axygen™
OTF50RPB
50 μL Green Filtered
Pack of 960 Each for $563.96
Case of 5 Pack for $2,819.80
Only null left
 
14-222-223
View Documents
Axygen™
OTF180RPB
180 μL Blue Filtered
Pack of 960 Each for $587.65
Case of 5 Pack for $2,938.25
Only null left
 
14-222-221
View Documents
Axygen™
OT200R
200 μL Blue Non-filtered
Pack of 960 Each for $419.09
Case of 5 Pack for $2,095.45
Only null left
 
14-221-901
View Documents
Axygen™
OTF100RPB
100 μL Gray Filtered
Pack of 960 Each for $547.22
Case of 5 Pack for $2,736.10
Only null left
 
14-223-312
View Documents
Axygen™
OT100R
100 μL Gray Non-filtered
Case of 4800 Each for $2,248.04
Only null left
 
Description

Description

  • Tip format: 96 tip
  • Tip volume range: 50 μL to 200 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
  • Compatible heads: 8- or 96-channel standard volume
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene
Specifications

Specifications

Non-sterile
Rack
SBS Rack
Dynamic Devices Oasis
Clear
Automation, Round
50 Racks/Cs.
Oasis
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'