missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Oasis™ Robotic Tips

Catalog No. 14222220
Change view
Click to view available options
Volume (Metric):
50 μL
100 μL
180 μL
200 μL
Rack Color:
Blue
Gray
Green
Filtered:
Filtered
Non-filtered
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Rack Color Filtered
14222220 50 μL Green Filtered
14222223 180 μL Blue Filtered
14222221 200 μL Blue Non-filtered
14221901 100 μL Gray Filtered
14223312 100 μL Gray Non-filtered
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14222220 Supplier Axygen™ Supplier No. OTF50RPB

For use with Oasis automated liquid handling systems

  • Tip format: 96 tip
  • Tip volume range: 50 μL to 200 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
  • Compatible heads: 8- or 96-channel standard volume
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene

Specifications

Volume (Metric) 50 μL
Filtered Filtered
Sterility Non-sterile
Color Clear
Rack Color Green
Clean Claims DNase, RNase Free
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Dynamic Devices Oasis
Product Line Oasis
Material Polypropylene
Show More Show Less