missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Oasis™ Robotic Tips

Catalog No. 14223312
Change view
Click to view available options
Volume (Metric):
50 μL
100 μL
180 μL
200 μL
Rack Color:
Blue
Gray
Green
Filtered:
Filtered
Non-filtered
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Rack Color Filtered
14223312 100 μL Gray Non-filtered
14222220 50 μL Green Filtered
14222223 180 μL Blue Filtered
14222221 200 μL Blue Non-filtered
14221901 100 μL Gray Filtered
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14223312 Supplier Axygen™ Supplier No. OT100R

For use with Oasis automated liquid handling systems

  • Tip format: 96 tip
  • Tip volume range: 50 μL to 200 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
  • Compatible heads: 8- or 96-channel standard volume
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene

Specifications

Volume (Metric) 100 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Rack Color Gray
Clean Claims DNase, RNase Free
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Dynamic Devices Oasis
Product Line Oasis
Material Polyethylene
Show More Show Less