missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Microplate Sealing Film and Tapes

Catalog No. 14222344
Change view
Click to view available options
Material:
Aluminum
Polyester
Polyolefin
Rayon
Product Type:
AxySeal Sealing Film
Breathable Sealing Film
CyclerSeal Sealing Film
Sealing Film
UltraClear Sealing Film
Dimensions (L x W):
133.3 x 79.2 mm
Not Available
135.5 x 80 mm
146 x 79.2 mm
146 x 79.6 mm
153 x 82.3 mm
Sterility:
Non-sterile
Sterile
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Product Type Dimensions (L x W) Sterility
14222344 Polyester AxySeal Sealing Film 146 x 79.6 mm Non-sterile
50400316 Rayon Breathable Sealing Film 153 x 82.3 mm Non-sterile
14222044 Rayon Breathable Sealing Film 153 x 82.3 mm Sterile
14222342 Aluminum Sealing Film 146 x 79.2 mm Non-sterile
14222343 Aluminum Sealing Film 133.3 x 79.2 mm Non-sterile
14222346 Polyester AxySeal Sealing Film 146 x 79.6 mm Sterile
14222347 Polyester CyclerSeal Sealing Film 135.5 x 80 mm Non-sterile
14222873 Polyolefin UltraClear Sealing Film Not Available Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 14222344 Supplier Axygen™ Supplier No. PCRSP
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Offer great economy for disposable ease.

For applications ranging from PCR and Real-Time PCR to ELISA and cell culture, Axygen films are an easy and cost-effective way to seal plates and enhance automation. Used to seal multi-well microplates. Sealing film temperature ranges from -40°C to +104°C.

Breathable

  • Allow effective gas exchange for cellular and bacterial cultivation — while preventing contamination
  • Seal polypropylene and polystyrene culture plates, 96- and 384-well plates including other assay plates
Aluminum
  • Provides exceptional seal on multi-well plates with varied temperatures and reagents
  • With compression mats, films and tapes seal during thermocycling
  • Ideal for light-sensitive samples and its uniformly applied adhesive provides optimum well sealing
  • Pierceable, PCR-AS-200
CyclerSeal
  • Exceptional seal across entire plate safeguards samples during transport or storage
  • When used with Axygen's compression mat, this polyester film eliminates well-to-well contamination and cross-over in PCR applications
AxySeal
  • Polyester-based with uniformly and consistently applied acrylic adhesive to eliminate the edge effect in sensitive ELISA assays
  • Suitable for sealing tissue culture plates, short-term storage and incubation, transport and the containment of biohazardous solutions
  • Available in both nonsterile and sterile packaging

Specifications

Material Polyester
Sterility Non-sterile
Length (English) 5.74 in.
Length (Metric) 146 mm
No. per Case 500/Cs.
No. per Pack 100/Pk.
Dimensions (L x W) 146 x 79.6 mm
Width (English) 3.13 in.
Width (Metric) 79.6 mm
For Use With (Application) Suitable for sealing tissue culture plates, for short term storage and incubation, for transport and the containment of biohazardous solutions
Product Type AxySeal Sealing Film
Adhesive Acrylic
Thickness 80 μm
Temperature Range (Metric) -80°C to +104°C
Show More Show Less