missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Axiom™ Strawberry FanaSNP 50k Genotyping Array, mini-96 format

Numéro de catalogue. 551042
Modifier l'affichage
Click to view available options
Quantité:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
551042 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 551042 Fournisseur Applied Biosystems™ Code fournisseur 551042

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

The Axiom FanaSNP Strawberry 50K Genotyping Array offers genome-wide coverage of polymorphic SNPs across the cultivated and wild octoploid strawberry species.

  • Array was designed by the University of California, Davis Strawberry Breeding Program in collaboration with Affymetrix Expert Design Program (Hardigan et al 2020)
  • The array offers genome-wide coverage of polymorphic SNPs across the cultivated and wild octoploid strawberry species
  • The Axiom FanaSNP Strawberry 50K Genotyping Array is also available in 384-HT format (Cat. Number 551041)

Array Content

  • Comprehensive content
    • 850,000 SNPs subgenome specific SNPs from diverse octoploid strawberry accessions from around the globe
    • Collectively, 446,644 of 850,000 marker probes produced QC-passing polymorphic SNP genotype clusters showing disomic (allopolyploid) segregation
    • The complete set of 446,644 validated probes is made available for public use
  • Production scale array also design
    • The most performant set of 49,483 SNPs was tested and validated for production activities
    • 5,809 SNPs from the iStraw 35K array were maintained for connectivity to previous platforms
  • Tied to multiple reference genomes
    • SNP positions ascertained by WGS alignment to the `Camarosa` v1 reference genome (Edger et al 2019) and have since been updated to the `UCD Royal Royce` (FaRR1) reference genome (Hardigan et al 2022)
  • Global represented and validated
    • Variants were discovered by WGS resequencing of 47 x ananassa, 24 F. chiloensis, and 22 F. virginiana individuals
    • Subgenome-specific marker performance variants were validated in a genetically diverse sample of 384 octoploid strawberry accessions

Applications

  • Molecular genetics research and inference
    • QTL discovery and Genome-wide association
    • Kinship and parentage analysis
  • Genomic and molecular breeding
    • Genomic prediction and selection
    • Marker-assisted selection
  • Release of new cultivars
    • Accelerate and increase efficiency of cultivar development
    • Identify the presence of important markers in existing strawberry germplasm
    • Use genetic information to identify germplasm that will perform well in specific growing regions
  • Intellectual property protection and verification
    • Clonal source and purity confirmation
    • Validation of parent-offspring relationships

Required Products

  • GeneChip Command Console Software
  • GeneTitan Multi-Channel Instrument
  • Axiom Analysis Suite Software
  • Axiom Long Format Export Tool (AxLE)

Spécifications

Array Genotyping
Nombre de matrices 96 arrays
Format Mini 96-array Plate
Espèces Strawberry
Quantité 1 plate

For Research Use Only. Not for use in diagnostic procedures.