missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axiom™ BreedWheat 35K Genotyping Array (mini-96 format)
Spécifications
Spécifications
| Format | Mini 96-array Plate |
| Contenu et stockage | One plate containing 96 arrays |
| Quantité | 1 plate |
For Research Use Only. Not for use in diagnostic procedures.