missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Avidin, NeutrAvidin™, Alexa Fluor™ 350 conjugué
Description
Our Alexa Fluor 350 conjugate of NeutrAvidin biotin-binding protein – a form of avidin that has been processed to remove carbohydrate and to lower its isoelectric point – can substantially decrease background due to nonspecific binding. The method used to deglycosylate the avidin retains its specific binding. This bright, blue-fluorescent reagent (absorption/emission maxima ∼346/442 nm) is ideal for detecting biotinylated targets in fluorescence microscopy and flow cytometry applications.
Spécifications
Spécifications
| Conjugué | Alexa Fluor 350 |
| Quantité | 1 mg |
| Contenu et stockage | À conserver au congélateur (de -5 à -30 °C) et à protéger de la lumière. |
| Shipping condition (Condition d'expédition) | Température ambiante |
| Gamme de produit | Alexa Fluor, NeutrAvidin |
| Étiquette ou colorant | Alexa Fluor Colorants |
Usage exclusivement réservé à la recherche. Ne pas utiliser dans des procédures de diagnostic.