Learn More
Invitrogen™ eBioscience™ Avidin HRP
Commonly used as a second step for detection of biotinylated antibodies by ELISA and western blot. Avidin binds to biotin with high affinity. [Note: Sodium azide is an inhibitor of horseradish peroxidase enzymatic activity and should not be added to the AV-HRP diluent.]
Supplier:  Invitrogen™ 18410051
 View more versions of this product
Catalog No. 501128941
Only null left
Explore available promotions
Description
Description
Avidin binds to biotin with high affinity. Because sodium azide is an inhibitor of horseradish peroxidase enzymatic activity, it should not be added to the AV-HRP diluent.
Specifications
Specifications
1 mL
Liquid
HRP (Horseradish Peroxidase)
Biotin
eBioscience
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.