missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Avidin, FITC conjugate

Catalog No. A821
Encompass
Change view
Click to view available options
Packaging:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
A821 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A821 Supplier Invitrogen™ Supplier No. A821

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Avidin, FITC conjugate

Our avidin, fluorescein conjugate has excitation and emission maxima ∼494/518. Avidin is a glycosylated biotin-binding protein that has a basic isoelectric point and is widely used to detect biotinylated probes.

Specifications

Conjugate FITC (Fluorescein)
Quantity 5 mg
Packaging 1 Each
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature
Product Type Avidin

For Research Use Only. Not for use in diagnostic procedures.