Learn More
FEATHER Autopsy Blade Handle F-100
Catalog No. 08923
Change view
Click to view available options
Quantity:
1 Pk.
Catalog No. Quantity
08923 1 Pk.
1 options
Catalog No. 08923 Supplier FEATHER Supplier No. F100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
FEATHER Autopsy Blade and Handle have superior sharpness and cutting performance compared to conventional knives. The handle is autoclavable and features excellent chemical resistance. The same handle can be used for brain dissection and other purposes simply by exchanging the blade.
  • Made in Japan
Specifications
Blade Material Special Resin (Autoclavable)
Product Type F-100
For Use With (Equipment) FEATHER Autopsy Blades No. 100, No. 170 and Brain Blade No. 325
Quantity 1 Pk.