missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms

Numéro de catalogue. 21402330
Modifier l'affichage
Click to view available options
Volume (Metric):
10 μL
20 μL
50 μL
175 μL
200 μL
1000 μL
Sterility:
Non-sterile
Sterile
Tip Style:
Automation
Conductive
Conductive, Wide Bore
Quantity:
96 Tips/Insert, 10 Inserts/Pk.
96 Tips/Tray, 50 Trays/Case
33 product options available for selection
Product selection table with 33 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Volume (Metric) Sterility Tip Style Quantity
21402330 10 μL Sterile Automation 96 Tips/Insert, 10 Inserts/Pk.
21402318 20 μL Sterile Conductive 96 Tips/Insert, 10 Inserts/Pk.
21402314 20 μL Non-sterile Conductive 96 Tips/Insert, 10 Inserts/Pk.
21402186 20 μL Non-sterile Conductive 96 Tips/Tray, 50 Trays/Case
21402331 20 μL Sterile Automation 96 Tips/Insert, 10 Inserts/Pk.
21402332 20 μL Non-sterile Automation 96 Tips/Insert, 10 Inserts/Pk.
21402323 50 μL Sterile Conductive 96 Tips/Insert, 10 Inserts/Pk.
21402333 50 μL Sterile Automation 96 Tips/Insert, 10 Inserts/Pk.
21402319 50 μL Sterile Conductive 96 Tips/Insert, 10 Inserts/Pk.
21402315 50 μL Non-sterile Conductive 96 Tips/Insert, 10 Inserts/Pk.
21402335 50 μL Sterile Automation 96 Tips/Insert, 10 Inserts/Pk.
21402334 50 μL Non-sterile Automation 96 Tips/Insert, 10 Inserts/Pk.
21402320 175 μL Sterile Conductive 96 Tips/Insert, 10 Inserts/Pk.
21402243 175 μL Sterile Conductive, Wide Bore 96 Tips/Insert, 10 Inserts/Pk.
21402336 175 μL Sterile Automation 96 Tips/Insert, 10 Inserts/Pk.
14754330 175 μL Non-sterile Conductive 96 Tips/Tray, 50 Trays/Case
21402316 200 μL Sterile Conductive 96 Tips/Insert, 10 Inserts/Pk.
21402312 200 μL Non-sterile Conductive 96 Tips/Insert, 10 Inserts/Pk.
21402188 200 μL Non-sterile Conductive 96 Tips/Tray, 50 Trays/Case
21402337 200 μL Sterile Automation 96 Tips/Insert, 10 Inserts/Pk.
21402338 200 μL Non-sterile Automation 96 Tips/Insert, 10 Inserts/Pk.
21402321 1000 μL Sterile Conductive 96 Tips/Insert, 10 Inserts/Pk.
21402246 1000 μL Sterile Conductive 96 Tips/Insert, 10 Inserts/Pk.
02681061 1000 μL Non-sterile Conductive 96 Tips/Tray, 50 Trays/Case
21402339 1000 μL Sterile Automation 96 Tips/Insert, 10 Inserts/Pk.
21402317 1000 μL Sterile Conductive 96 Tips/Insert, 10 Inserts/Pk.
21402247 1000 μL Sterile Conductive 96 Tips/Insert, 10 Inserts/Pk.
21402313 1000 μL Non-sterile Conductive 96 Tips/Insert, 10 Inserts/Pk.
14754466 1000 μL Non-sterile Conductive 96 Tips/Insert, 10 Inserts/Pk.
21402189 1000 μL Non-sterile Conductive 96 Tips/Tray, 50 Trays/Case
21402340 1000 μL Sterile Automation 96 Tips/Insert, 10 Inserts/Pk.
21402341 1000 μL Non-sterile Automation 96 Tips/Insert, 10 Inserts/Pk.
21402322 10 μL Sterile Conductive 96 Tips/Insert, 10 Inserts/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 33 options available.
33 options
Numéro de catalogue. 21402330
Il en reste null

Benefit from Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms. They increase pipetting performance through the superior tip straightness and low %CV of these pipette tips.

Get accuracy and precision with Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance.

  • Available as standard, filtered and/or liquid sensing
  • Certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
  • Lot-tested during production on actual workstation for which they are created
  • Fully conductive, providing increased pipetting performance
  • Low percentage coefficient of variation (%CV)

Spécifications

Volume (Metric) 10 μL
Filtered Filtered
Sterility Sterile
Color Clear
Rack Color Gray
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001 and ISO 13485
Clean Claims Free of RNase, DNase, DNA, ATP, and endotoxin
Format Reload Deck
Tip Style Automation
Packaging Blister Pack
Quantity 96 Tips/Insert, 10 Inserts/Pk.
For Use With (Equipment) Tecan Systems with LiHa Head
Product Line Tecan
Fit for Purpose Sterile and PCR Ready
Afficher plus de résultats Afficher moins de résultats