missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Automation Tips for PerkinElmer™/Packard™ Robotic Liquid Handling Systems with 96- and 384-Channel MDT Dispense Heads

Catalog No. 361024378
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
30 μL
40 μL
50 μL
180 μL
235 μL
Autoclavable:
Autoclavable
Not Autoclavable
Filtered:
Filtered
Non-filtered
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Autoclavable Filtered Fit for Purpose
361024378 180 μL Not Autoclavable Filtered Sterile and PCR Ready
21402635 10 μL Not Autoclavable Filtered Sterile and PCR Ready
21402633 20 μL Not Autoclavable Non-filtered Sterile
21402634 20 μL Autoclavable Non-filtered Certified Quality
21402632 40 μL Not Autoclavable Filtered Sterile and PCR Ready
21402630 50 μL Not Autoclavable Non-filtered Sterile
21402631 50 μL Autoclavable Non-filtered Certified Quality
2140325 235 μL Not Autoclavable Non-filtered Sterile
2140326 235 μL Autoclavable Non-filtered Certified Quality
21931261 30 μL Not Autoclavable Non-filtered Sterile
14754331 50 μL Not Autoclavable Non-filtered Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 361024378 Supplier Thermo Scientific™ Supplier No. 929021

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Increase pipetting effectiveness with these automation tips, achieving accurate and precise performance due to superior tip straightness and a low coefficient of variation.

Thermo Scientific™ Automation Tips for Perkin Elmer™/Packard™ Robotic Liquid Handling Systems with Varispan™ Dispense Arms, which undergo a rigorous 15-point quality control program to ensure consistency. This leads to accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created and the sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.

Features:

  • Available as sterile and filtered
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
  • Lot-tested during production on actual workstation for which they are created
  • Low percentage coefficient of variation (%CV)

Specifications

Volume (Metric) 180 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001 and ISO 13485
Clean Claims Free of RNase, DNase, DNA, ATP, and endotoxin
Format Rack
Tip Style Automation
Packaging 96 tips/rack, 10 racks/pack, 5 packs/case
Quantity 4800 tips
For Use With (Equipment) Packard Liquid Handling System, PerkinElmer
Product Line PerkinElmer/Packard
Fit for Purpose Sterile and PCR Ready
Show More Show Less