Learn More
BeLLCo Glass Manufactured from medium wall Borosilicate Glass Type 1 Class A, with permanent graduations designed for DNA/RNA protocols. Includes high temperature red PBT cap with PTFE faced silicone liner. Replacement cap 5639-90045
Numéro de catalogue. 501946511
Encompass
Manufactured from medium wall Borosilicate Glass Type 1 Class A, with permanent graduations designed for DNA/RNA protocols. Includes high temperature red PBT cap with PTFE faced silicone liner. Replacement cap 5639-90045
Numéro de catalogue. 501946511 Fournisseur BeLLCo Glass Code fournisseur 791035010
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Manufactured from medium wall Borosilicate Glass Type 1 Class A, with permanent graduations designed for DNA/RNA protocols. Includes high temperature red PBT cap with PTFE faced silicone liner. Replacement cap 5639-90045