missing translation for 'onlineSavingsMsg'
Learn More

Aushon Chemiluminescent Soluble Human Cytokine Receptor Array Kits

Catalog No. 07F801FA
Encompass
Change view
Click to view available options
Quantity:
1 Plex
2 Plex
4 Plex
5 Plex
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07F801FA 5 Plex
070011FA 1 Plex
070061FA 2 Plex
070781FA 4 Plex
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07F801FA Supplier Aushon Biosystems Inc Supplier No. 07F801FA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For simultaneous measurement of combinations of soluble cytokine receptors

Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate

Specifications

Reactivity Human
For Use With (Application) For simultaneous measurement of combinations of soluble Cytokine receptors
Quantity 5 Plex