Learn More
Aushon Chemiluminescent Rat Cytokine Array Kits
Catalog No. 233FF1FA
Encompass
Change view
Click to view available options
Detects:
Cytokine 1
Cytokine 2
Cytokine 3
Cytokine 4
Cytokine 5
Cytokine 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detects
233FF1FA Cytokine 1
232921FA Cytokine 5
233F61FA Cytokine 2
232941FA Cytokine 6
232B01FA Cytokine 4
233961FA Cytokine 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 233FF1FA Supplier Aushon Biosystems Inc Supplier No. 233FF1FA
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For simultaneous measurement of combinations of cytokines
Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles
Specifications
Detects Cytokine 1
For Use With (Application) For simultaneous measurement of combinations of Cytokines
Product Type Cytokine Array Kits
Quantity 10 Plex