missing translation for 'onlineSavingsMsg'
Learn More

Aushon Chemiluminescent Human Growth Hormone 1 Array Kits

Catalog No. 110011FA
Encompass
Change view
Click to view available options
Quantity:
1 Plex
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
110011FA 1 Plex
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 110011FA Supplier Aushon Biosystems Inc Supplier No. 110011FA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For simultaneous measurement of combinations of growth hormones

Using a CCD imaging system and array software, kits allow you to quantify multiple proteins per well in a 96-well plate, generating chemokine profiles

Specifications

Reactivity Human
For Use With (Application) For simultaneous measurement of combinations of growth hormones
Quantity 1 Plex