missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Auramine, Norme analytique, ≥97,0% (HPLC), MilliporeSigma™ Supelco™
Spécifications
Spécifications
| Packaging | Bouteille en verre sans fond avec cône fusionné |
| CAS | 492-80-8 |
| Physical Form | Sympa |
| Linear Formula | C17H21N3 |
| MDL Number | MFCD00036224 |
| Synonym | 4,4 '-Dimethylaminobenzophenone imine; Solvent Yellow 34 |
| Quantity | 25 mg |
| Formula Weight | 267.37 |
| Percent Purity | ≥97.0% (HPLC) |
| Grade | Norme analytique |