missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT and CytPix Level Sensor Cable

Catalog No. 502723658
Change view
Click to view available options
Quantity:
1 cable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
502723658 1 cable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 502723658 Supplier Invitrogen™ Supplier No. 100087973CST

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This is a replacement level sensor cable for the focusing fluid and waste bottles of the Attune NxT Acoustic Focusing Cytometer

  • This is a replacement level sensor cable for the focusing fluid and waste bottles of the Attune NxT Acoustic Focusing Cytometer.
  • It can be replaced easily without replacing the entire bottle assembly if the sensor connector is broken.

Specifications

Format Sensor cable
Quantity 1 cable
Content And Storage 1 Cable
Product Type Flow Cytometer Accessory
Shipping Condition Room Temperature
For Use With (Equipment) Attune NxT
Product Line FPL