Learn More
Invitrogen™ Attune™ NxT Flow Cytometer, blue

Description
With the Attune™ NxT Acoustic Focusing Cytometer you get:
Flexible design that accommodates changing needs, budget, or laboratory space
The instrument’s modular design accommodates the most common fluorophores and fluorescent proteins used in flow cytometry enabling alignment with previous experimental results and accommodating future needs through simple upgrades. The small footprint of the instrument requires minimal bench space, even allowing use in a cell culture hood.
Acoustic focusing improves efficiencies compared to traditional flow cytometers
Traditional flow cytometers rely on a large volume of fluid to focus cells prior to analysis, resulting in extensive sample preparation prior to analysis and introducing the possibility of error due to artifacts created during the sample preparation process. With the Attune™ NxT Acoustic Focusing Cytometer, blood samples can be run without being manipulated using the No-Wash No-Lyse blood cell assay. The instrument also offers flexibility in the sample concentration that can be analyzed.
Acoustic focusing enables:
Learn more about the Attune NxT cytometer and see all available configurations >
Learn more about how acoustic focusing works >
Specifications
Specifications
| Content And Storage | • Attune NxT Flow Cytometer • Minitower Windows 10 64-bit computer with Attune Cytometric Software • Power cord with adapters • USB cable • Attune Shutdown Solution • Attune Wash Solution • Attune Focusing Fluid • Attune NxT Flow Cell Cleaning Solution • Attune Performance Tracking Beads • Quick reference card |
| Format | Tube(s) |
| Laser Features | Blue Excitation Laser (488 nm) |
| Frequency | 50/60 Hz |
| Product Line | Attune |
| For Use With (Equipment) | Attune™ NxT Acoustic Focusing Cytometer |
| Shipping Condition | Room Temperature |
| Quantity | 1 each |
Frequently Asked Questions (FAQs)
To minimize the number of failed performance tests on your Attune NxT Flow Cytometer, we suggest the following:
• Make sure to carry out daily, weekly, and monthly maintenance procedures (pages 8-14 of the Maintenance and Troubleshooting Guide, linked below).
• Always run 3 startup cycles and 2 Rinse procedures (pages 46-47 of the Maintenance and Troubleshooting Guide linked below).
• If the performance test fails, carry out the following priming sequence: run 3 startup cycles, 2 de-bubble cycles, and 5 rinse procedures, followed by running the SIP sanitize function.
Please review the Attune Flow Cytometry Maintenance and Troubleshooting Guide for more information about performing routine maintenance procedures and troubleshooting.
Yes. The Attune Nxt Flow Cytometer can detect down to 0.2 µm and using a special configuration, down to 0.1 µm. Optimize your settings using Flow Cytometry Sub-micron Particle Size Reference Kit, Cat. No. F13839.
Yes. You would need to purchase Attune NxT Custom Filter Holder Kit, Cat. No. A27784 to make your own custom emission or dichroic filters in order to customize the optical configuration of the instrument. Each kit contains sufficient parts to make 2 emission filters and 2 dichroic filters. The kit includes instructions for attaching the filter to the filter blade, but does not include the glass filter. Other accessories to optimize for special applications include Cat No. 100083194 Attune NxT Small Particle Side-Scatter Filter, Cat. No. 100083194 and Attune NxT Blocker Bar Conversion Kit, Cat No. A35966.
Yes. There are 6 lasers to choose from (blue, green, yellow, red, violet, and violet 6). You can start with one laser and at any time upgrade the same instrument with up to 4 lasers to utilize a maximum of 14 detection channels. Using the Violet 6 laser along with the blue, red, and yellow lasers, one can detect up to 16 channels.
Because of the acoustic-assisted hydrodynamic technology, the Attune NxT Flow Cytometer uses only 1/10th the amount of fluids, and is 10X faster and less sensitive to clogging compared to standard flow cytometers. You can set the flow rate from 12.5-1,000 µL/min and collect up to 65,000 events/sec using from 20 µL to 4 mL of sample volume. These unique properties allow for rare event analysis, as well standard flow analysis.
For Research Use Only. Not for use in diagnostic procedures.