missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT Flow Cytometer, blue/red/green

Catalog No. A28997
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28997 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A28997 Supplier Invitrogen™ Supplier No. A28997

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed and developed to remove common barriers associated with flow cytometry, the evolutionary capabilities of the Attune NxT Flow Cytometer

  • The 2020 Attune NxT Flow Cytometer has expanded functionalities to drive your research forward. Designed and developed to remove common barriers associated with flow cytometry, the evolutionary capabilities of the Attune NxT Flow Cytometer offer adaptable optical configuration options so you can get the most out of your multicolor analysis.
  • New CytKick and CytKick Max autosampler—more efficiency for high-throughput assays
  • New 21 CFR part 11 compliant software for regulated laboratories on Windows 10 operating system
  • Improved workflow for run protocol and instrument settings management
  • The innovative combination of acoustic focusing technology coupled with traditional hydrodynamic focusing results in acquisition speeds up to 10X faster than other flow cytometers. This technology reduces the time needed to obtain results.
  • True 35,000 event/sec (+10% coincidence rates)
  •  Maximum electronic speed of 65,000 events/sec
  •  Analyze small or limited samples faster than ever
  • Efficiently run traditionally long assays
  • The Attune NxT Flow Cytometer is ideal for immunophenotyping and signaling studies, cell cycle analysis, detection of rare events, stem cell analysis, cancer and apoptosis studies, microbiological assays, and more.
  • The instrument’s modular design provides the flexibility to meet future capability needs with a variety of upgrade options. Purchase what you need now and easily upgrade later. The small footprint of the instrument requires minimal bench space, even allowing use in a cell culture hood.
  • Flexibility—1 to 4 lasers, 14 colors, and 16 parameters
  • Customizable band pass filters for expanded fluorochrome choices
  • The Attune NxT Flow Cytometer easily runs most samples including cardiomyocytes. The system is designed with a clog-resistant syringe-driven system to push the sample with a force of up to 75 psi.
  • Flexibility in sample concentrations and sample types
  • Elimination of time-consuming concentration steps with No-Wash, No-Lyse Filter Kit
  • Analysis of dilute samples, including CSF, stem cells, mouse bone marrow, mouse blood, and thin needle aspirates
  • Acquisition of a high volume of samples in minimal time

Specifications

Content And Storage • Attune NxT Flow Cytometer
• Minitower Windows 10 64-bit computer with Attune Cytometric Software
• Power cord with adapters
• USB cable
• Attune Shutdown Solution
• Attune Wash Solution
• Attune Focusing Fluid
• Attune NxT Flow Cell Cleaning Solution
• Attune Performance Tracking Beads
• Quick reference card

Format Tube
Laser Features 488 nm, 637 nm, 532 nm
Product Line Attune
For Use With (Equipment) Attune™ Acoustic Focusing Cytometer
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Quantity 1 each
My Attune NxT Flow Cytometer performance test keeps failing. What steps can I take to minimize the number of failed performance tests?

To minimize the number of failed performance tests on your Attune NxT Flow Cytometer, we suggest the following:

•   Make sure to carry out daily, weekly, and monthly maintenance procedures (pages 8-14 of the Maintenance and Troubleshooting Guide, linked below).
•   Always run 3 startup cycles and 2 Rinse procedures (pages 46-47 of the Maintenance and Troubleshooting Guide linked below).
•   If the performance test fails, carry out the following priming sequence: run 3 startup cycles, 2 de-bubble cycles, and 5 rinse procedures, followed by running the SIP sanitize function.

Please review the Attune Flow Cytometry Maintenance and Troubleshooting Guide for more information about performing routine maintenance procedures and troubleshooting.

Can I use the Attune Nxt Flow Cytometer to detect bacteria?

Yes. The Attune Nxt Flow Cytometer can detect down to 0.2 µm and using a special configuration, down to 0.1 µm. Optimize your settings using Flow Cytometry Sub-micron Particle Size Reference Kit, Cat. No. F13839.

Of all the Attune NxT filters you offer, I don't see one that matches our application. Can I use a filter other than what you offer?

Yes. You would need to purchase Attune NxT Custom Filter Holder Kit, Cat. No. A27784 to make your own custom emission or dichroic filters in order to customize the optical configuration of the instrument. Each kit contains sufficient parts to make 2 emission filters and 2 dichroic filters. The kit includes instructions for attaching the filter to the filter blade, but does not include the glass filter. Other accessories to optimize for special applications include Cat No. 100083194 Attune NxT Small Particle Side-Scatter Filter, Cat. No. 100083194 and Attune NxT Blocker Bar Conversion Kit, Cat No. A35966.

With the Attune Nxt Flow Cytometer, if I wish to only start with a single laser, can I expand to more lasers and channels?

Yes. There are 6 lasers to choose from (blue, green, yellow, red, violet, and violet 6). You can start with one laser and at any time upgrade the same instrument with up to 4 lasers to utilize a maximum of 14 detection channels. Using the Violet 6 laser along with the blue, red, and yellow lasers, one can detect up to 16 channels.

What makes the Attune Nxt Flow Cytometer better than standard flow cytometers?

Because of the acoustic-assisted hydrodynamic technology, the Attune NxT Flow Cytometer uses only 1/10th the amount of fluids, and is 10X faster and less sensitive to clogging compared to standard flow cytometers. You can set the flow rate from 12.5-1,000 µL/min and collect up to 65,000 events/sec using from 20 µL to 4 mL of sample volume. These unique properties allow for rare event analysis, as well standard flow analysis.

How often should I change filters for the Attune NxT Acoustic Focusing Cytometer?

Filters are recommended for replacement once every three months, unless performing small particle detection when once-a-month replacement is recommended.

I'd like to use the Flow Cytometry Panel Builder but will be purchasing antibodies from a different vendor than Thermo Fisher Scientific. Is this possible?

Step 2 allows for antibodies you have already purchased or are from a vendor outside Thermo Fisher Scientific. If you are undecided about which format you want for something, you can add the antigen name in the "Antibodies you need" section and then choose a placeholder fluorochrome in the following step. Both will help you keep track of channels that are reserved for reagents that you do not plan to buy from Thermo Fisher Scientific.

Can I export the spreadsheet from the Flow Cytometry Panel Builder for later use?

There are both PDF (printable) and CSV (spreadsheet) exports available in the summary step for you to take away for reference and ordering purposes.

In the Flow Cytometry Panel Builder tool, I can't find my flow cytometer in the dropdown. Can I build one?

Yes, you can define a new cytometer using the "Enter your cytometer manually" link below the cytometer dropdown menu.

I would like to use the Flow Cytometry Panel Builder but my flow cytometer does not use a standard configuration. How can I make changes to one in the dropdown?

Select a cytometer from the dropdown menu and then select the "Edit cytometer settings" link to modify the configuration as needed.

In the Flow Cytometry Panel Builder tool, what do the colorful dots indicate in the "Fluorochromes" step?

Each dot indicates a different fluorochrome that Thermo Fisher Scientific has available in each channel of the cytometer you have chosen.

How do I share my panel generated using the Flow Cytometry Panel Builder with my lab manager?

You may export the spreadsheet found on step 5 and share it.

In the Flow Cytometry Panel Builder, my marker isn't showing up under step 2 of antigen, but I see it in the catalog. How can I remedy this?

Select your target species first, and then type your antigen. A drop-down list will appear. Select the antigen from the list and click on "Confirm selections." If you have an antibody that is not on the list, in the "Antibodies you already have" section, you can enter a name and select "use this name anyway."

In the Flow Cytometry Panel Builder tool, can I go back a step without losing the work I did?

Yes, the previous work is saved. If you plan to leave the Panel Builder and come back later, be sure to select "Save" in the lower right corner to save your panel.

Is an isotype control a must for flow cytometry analysis?

No. Many users are using unstained cells in combination with FMO controls to identify their positive populations.

I am using the Attune Nxt Acoustic Focusing Cytometer. Why is my event rate so low?

There are several things that can cause a low event rate:

-The system may be clogged
-The threshold level may be set too high
-The PMT voltage for the threshold parameter may be set too low
-The sample may be too dilute or not adequately mixed
-The sample syringe may be loose

I am using the Attune Nxt Acoustic Focusing Cytometer. Why is my event rate so high?

There are several things that can cause a high event rate:

-There may be an air bubble in the flow cell
-The threshold level may be set too low
-The PMT voltage for the threshold parameter may be set too high
-The sample may be too concentrated
-The sample flow rate may be too high
-The sample may have a bacterial contamination

Where can I find the log file for my Attune Nxt instrument?

Open the Attune NxT software, open the main menu, and go to the “Instrument” tab. The option on the far left is “System Log”. Click on that and it will open the log file. At the bottom of the page there is a button labeled “Export”. Use this to export and save the log file.

I ordered a box of Attune Focusing Fluid (10L, A24904) and I cannot find the spigot. What should I do?

We recommend looking inside the box, possibly underneath the bag of buffer.

I am using the Attune Nxt Acoustic Focusing Cytometer and my data does not look like I expected. How can I get help with it?

Send an email to Cellular Analysis Technical Support at technical-molecularprobes@lifetech.com, including the words “flow*#148; or “Attune” in the subject line for proper routing of your email. Please describe the type of application you are doing, how you set up the experiment, and your results. It would be ideal if you could send us a sample of your data that we can review.

I have an Attune Nxt Acoustic Focusing Cytometer and it’s not working right. Whom do I contact?

If the issue is clearly an instrument problem, send an email to Instrument Services at instrumentservices@thermofisher.com with your instrument serial number. If the issue is software or application-related, or if you do not know the source of the problem, send an email to Cellular Analysis Technical Support at technical-molecularprobes@lifetech.com, including the words “flow*#148; or “Attune” in the subject line for proper routing of your email.

Can you help me design my antibody panel for immunophenotyping?

For help with designing panels for flow cytometry, see our new Molecular Probes Flow Cytometry Panel Design Tool (http://www.thermofisher.com/us/en/home/life-science/cell-analysis/flow-cytometry/flow-cytometry-panel-design-tool.html) The panel design tool can help you choose fluorescent antibody conjugates for your flow cytometry panel in a few easy steps: pick the antibody species reactivity, select up to 14 targets of interest (choices include viability dyes), and choose the lasers or fluorophores you want to view.

How many different parameters can you combine in one assay?

It depends on your instrument. People have done 17 colors (Perfetto SP, Chattopadhyay PK, and Roederer M (2004) Seventeen-colour flow cytometry: unraveling the immune system. Nat Rev Immunol 4:648–655 (http://www.ncbi.nlm.nih.gov/pubmed/15286731)) and more, but it takes a lot of planning and testing to make it work. The Attune NxT Acoustic Focusing Cytometer can run up to 14 colors plus forward and side scatter with a system equipped with all four lasers (violet, blue, yellow and red).

What is an FMO control in flow cytometry?

An FMO control (flow minus one) is a control in which you label cells or beads with every color in your panel, omitting one. Make one FMO control for each color in your panel. These controls are important for helping you properly set gates on your data.

What if I can't find the right isotype control for flow cytomtery?

The flow cytometry field is moving away from using isotype controls as they are not necessarily the most appropriate way to control for non-specific binding. Instead, they are using unstained cells to define the negative population, single stained controls to set compensation, and flow minus one (FMO) controls to set regions and gates.

You may want to consider whether using an isotype control is something you need to do. Here are some references you might want to look at:
-O'Gorman MR, Thomas J. (1999) Isotype controls-time to let go? Cytometry 38:78-80.

-Keeney M, Gratama JW, Chin-Yee IH et al. (1998) Isotype controls in the analysis of lymphocytes and CD34+ stem/progenitor cells by flow cytometry- time to let go! Cytometry 34:280-283.

-Hulspas R, O'Gorman MR, Wood BL et al. (2009) Considerations for the control of background fluorescence in clinical flow cytometry. Cytometry B Clin Cytom 76:355–364.

-Enumeration of Immunologically Defined Cell Populations by Flow Cytometry; Approved Guideline - Second Edition. Clinical and Laboratory Standards Institute, (CLSI). Document H42-A2 Volume 27 No.16, 2007.

-Clinical Flow Cytometric Analysis of Neoplastic Hematolymphoid Cells; Approved Guideline - Second Edition. Clinical and Laboratory Standards Institute, (CLSI). Document H43-A2 Volume 27 No. 11, 2007.

If you do wish to use isotype controls, we offer a wide variety of species and fluorophores, search our Web site under "isotype antibody control."

What is the Flow Cytometry Size Calibration Kit (Cat. No. F13838) used for?

Our Flow Cytometry Size Calibration Kit has non-fluorescent particle-size calibration standards that provide a simple, accurate way to determine cell sizes by flow cytometry. The kit contains six suspensions of highly uniform polystyrene microspheres with the following diameters: 1.0 µm, 2.0 µm, 4.0 µm, 6.0 µm, 10.0 µm and 15.0 µm. All of the microsphere suspensions are provided in convenient dropper vials. This kit looks at the scatter properties of non-fluorescent microspheres of known sizes to determine approximate cell sizes.

What is the smallest size that I can detect with the Attune NxT Acoustic Focusing Cytometer?

The smallest size that you can detect with the Attune NxT Acoustic Focusing Cytometer is 0.5 µm.

What is the Attune NxT Autosampler?

The Attune NxT Autosampler, an optional accessory for the Attune NxT Acoustic Focusing Cytometer, enables rapid processing of up to 384 samples. It has broad compatibility with different plate formats, both 96- and 384-well plates. It has an intelligent probe designed to minimize clogging and carryover (<0.5%) and to prevent damage to the instrument. It mixes by aspiration rather than shaking to ensure homogeneity of the sample and maintain cell viability. Is performs automated cleaning as part of the shutdown process of the Attune NxT Cytometer. It provides consistent data regardless of sampling method (tube vs. plate) and collection rate.

What are the advantages of acoustic-assisted hydrodynamic focusing in flow cytometry?

-Modular design - Multiple configurations available - field upgradable.
-Save time - 10X faster speeds with no loss in data quality.
-Simplified sample prep - No wash, no lyse options, non-clogging fluidics.
-Enables unique applications - Complex protocols on a broad range of cell types and samples.

How is the Attune NxT Acoustic Focusing Cytometer different from traditional flow cytometers?

With the option to be configured with up to 4 lasers and 14 colors for multi-parameter analysis the Attune NxT Acoustic Focusing Cytometer was designed as a modular system to fit most experimental needs and lab budgets. The novel design of the optical path helps ensure precise fixed alignment of four spatially separated lasers onto the sample stream enabling consistency in data over time, superior performance, and superior reliability. The instrument can be configured with up to 4 solid-state lasers (405 nm, 488 nm, 561 nm, and 637 nm) with flat top beam profiles.

The Attune NxT Flow Cytometer's acoustic focusing uses ultrasonic radiation pressure (> 2 MHz) to transport particles into the center of the sample stream. This pre-focused stream is then injected into the sheath stream, which supplies an additional hydrodynamic pressure to the sample. The combination of these two forces- termed acoustic-assisted hydrodynamic focusing-results in a narrow core stream and uniform laser illumination, regardless of the sample input rate. In traditional cytometers that rely solely on hydrodynamic focusing, the sample core widens to accommodate the increases in flow rate, which results in less uniform laser light illumination.

What is flow cytometry?

Cytometry is the measurement of physical or chemical characteristics of cells or particles. Flow cytometry measures these characteristics of cells or particles as they individually pass lasers in a flow cytometer instrument. Flow cytometry is performed on single cells, providing discrete measurements for each cell in the sample. It also provides a statistical distribution of the measured characteristics of the sample.

A flow cytometer is made up of three subsystems: fluidics, optics, and electronics. Fluidics moves the cells and introduces them for interrogation. Optics generates and collects the light signals. Electronics converts the optical signals to proportional electronic signals for computer analysis.