missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Attune™ Focusing Fluid (1X)
Attune™ Focusing Fluid (1X)
$50.50 - $257.00
Specifications
| Product Type | Focusing Fluid |
|---|---|
| For Use With (Equipment) | Attune™ Acoustic Focusing Cytometer |
| Product Line | Attune |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
501034959
|
N/A | 6 x 1 L |
Case of 6 Each for $206.28
|
|
|||||
|
4449791
|
N/A | 6 x 1 L |
Each for $143.00
|
|
|||||
501034964
|
N/A | 10 L |
Each for $228.70
|
|
|||||
|
A24904
|
N/A | 10 L |
Each for $145.00
|
|
|||||
|
4488621
|
N/A | 1 L |
Each for $50.50
|
|
|||||
501034960
|
N/A | 1 L |
Each for $80.42
|
|
|||||
|
J106627
|
N/A | 20 L |
Each for $257.00
|
|
|||||
Description
Attune™ Focusing Fluid is an optimized buffer solution that is used in the Attune™ Acoustic Focusing Cytometer for focusing and transportation of particles throughout the fluidics system.Maintains System Cleanliness
Attune™ Focusing Fluid is a sterile, buffered, azide-free support reagent that contains a surfactant that helps clean the fluidic lines of the Attune™ Acoustic Focusing Cytometer. The Attune™ Focusing Fluid was designed to minimize bubble formation and to prevent build-up of cellular material in the flow cell, resulting in less instrument clogging.
Convenient
The ready-to-use 1X focusing fluid was designed to eliminate the need for dilution prior to use, as well as access to a purified water system. The convenient 10 L packaging provides enough focusing fluid to operate for at least 10 days (the average focusing fluid consumption per day is less than 1 L) and has a spigot to to make filling of the onboard fluidics tanks easier.
High Quality
Attune™ Focusing Fluid is prepared, filtered, and packaged in a sterile environment to ensure the highest quality. Each batch is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer.
Specifications
| Focusing Fluid | |
| Attune™ Acoustic Focusing Cytometer | |
| Attune |
For Research Use Only. Not for use in diagnostic procedures.