missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Attune™ Focusing Fluid (1X)
Attune™ Focusing Fluid (1X)
Supplier: Life Sciences Group (Lsg) A24904
Description
Attune™ Focusing Fluid is an optimized buffer solution that is used in the Attune™ Acoustic Focusing Cytometer for focusing and transportation of particles throughout the fluidics system.Maintains System Cleanliness
Attune™ Focusing Fluid is a sterile, buffered, azide-free support reagent that contains a surfactant that helps clean the fluidic lines of the Attune™ Acoustic Focusing Cytometer. The Attune™ Focusing Fluid was designed to minimize bubble formation and to prevent build-up of cellular material in the flow cell, resulting in less instrument clogging.
Convenient
The ready-to-use 1X focusing fluid was designed to eliminate the need for dilution prior to use, as well as access to a purified water system. The convenient 10 L packaging provides enough focusing fluid to operate for at least 10 days (the average focusing fluid consumption per day is less than 1 L) and has a spigot to to make filling of the onboard fluidics tanks easier.
High Quality
Attune™ Focusing Fluid is prepared, filtered, and packaged in a sterile environment to ensure the highest quality. Each batch is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer.
Chemical Identifiers
| Focusing Fluid | |
| Attune™ Acoustic Focusing Cytometer | |
| Attune |
Specifications
| Attune | |
| Attune™ Acoustic Focusing Cytometer | |
| Focusing Fluid |
| 10 L | |
| Store at room temperature. |
For Research Use Only. Not for use in diagnostic procedures.