missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Atropic Acid | |
| 107°C | |
| 267°C | |
| MFCD00046531 | |
| 2-phenylacrylic acid, atropic acid, acrylic acid, 2-phenyl, 2-propenoic acid, 2-phenyl, unii-tw16ua35s0, benzeneacetic acid, .alpha.-methylene, .alpha.-phenyl acrylic acid, .alpha.-toluic acid, .alpha.-methylene, alpha-phenylacrylic acid, alpha-phenyl acrylic acid | |
| C=C(C1=CC=CC=C1)C(=O)O | |
| 148.161 | |
| 148.16 | |
| Crystalline Powder |
| 492-38-6 | |
| White-Yellow | |
| C9H8O2 | |
| 25 g | |
| ONPJWQSDZCGSQM-UHFFFAOYSA-N | |
| 2-phenylprop-2-enoic acid | |
| 68114 | |
| ≥93.0% (GC) |
Chemical Identifiers
| 492-38-6 | |
| 148.161 | |
| ONPJWQSDZCGSQM-UHFFFAOYSA-N | |
| 68114 | |
| C=C(C1=CC=CC=C1)C(=O)O |
| C9H8O2 | |
| MFCD00046531 | |
| 2-phenylacrylic acid, atropic acid, acrylic acid, 2-phenyl, 2-propenoic acid, 2-phenyl, unii-tw16ua35s0, benzeneacetic acid, .alpha.-methylene, .alpha.-phenyl acrylic acid, .alpha.-toluic acid, .alpha.-methylene, alpha-phenylacrylic acid, alpha-phenyl acrylic acid | |
| 2-phenylprop-2-enoic acid |
Safety and Handling
TSCA : No