accessibility menu, dialog, popup

UserName

OHOCH2

CAS RN 492-38-6

IUPAC Name: 2-phenylprop-2-enoic acid
Synonyms: atropic acid
Molecular Weight (g/mol): 148.16
Molecular Formula: C9H8O2
InChi Key: ONPJWQSDZCGSQM-UHFFFAOYSA-N
SMILES: OC(=O)C(=C)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)