missing translation for 'onlineSavingsMsg'
Learn More

Atropic Acid 93.0+%, TCI America™

Catalog No. p-7117876
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23905G 5 g
A239025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23905G Supplier TCI America Supplier No. A23905G
Only null left

Chemical Identifiers

CAS 492-38-6
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00046531
InChI Key ONPJWQSDZCGSQM-UHFFFAOYSA-N
Synonym 2-phenylacrylic acid, atropic acid, acrylic acid, 2-phenyl, 2-propenoic acid, 2-phenyl, unii-tw16ua35s0, benzeneacetic acid, .alpha.-methylene, .alpha.-phenyl acrylic acid, .alpha.-toluic acid, .alpha.-methylene, alpha-phenylacrylic acid, alpha-phenyl acrylic acid
PubChem CID 68114
IUPAC Name 2-phenylprop-2-enoic acid
SMILES C=C(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point 107°C
Color White-Yellow
Boiling Point 267°C
Quantity 5 g
Formula Weight 148.16
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Atropic Acid