missing translation for 'onlineSavingsMsg'
Learn More

Atrazine, MilliporeSigma™ Supelco™

Catalog No. 111020255
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020255 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020255 Supplier Merck Emd Millipore Supplier No. 49085

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Atrazine is a commonly used herbicide, which belongs to the triazine class of herbicides. It is generally used for weed control management in agricultural crops.

Agriculture and Environmental

Specifications

Packaging Ampule
CAS 1912-24-9
Physical Form Neat
Linear Formula C8H14 ClN5
MDL Number MFCD00041810
Synonym 2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine
Type Analytical Standard
Quantity 100 mg
Formula Weight 215.68
Grade Analytical Standard