missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATP13A2 Polyclonal Antibody

Description
Suggested positive control: human and mouse brain membrane prep lysate.
Neurodegenerative disorders such as Parkinson and Alzheimer disease cause motor and cognitive dysfunction and belong to a heterogeneous group of common and disabling disorders. ATP13A2, otherwise known as PARK9, is a neuronal P-type ATPase gene underlying an autosomal recessive form of early-onset parkinsonism with pyramidal degeneration and dementia. ATP13A2 protein is located in the membrane of these lysosomes and is formed most strongly in the brain, especially in the substantia nigra, a brain region which is known to play a central role in Parkinson's disease.
Spécifications
Spécifications
| Antigène | ATP13A2 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.75 mg/mL |
| Conjugué | Unconjugated |
| Formule | TBS with 0.05% sodium azide |
| Gène | ATP13A2 |
| Numéro d’acquisition du gène | Q9CTG6, Q9NQ11 |
| Alias du gène | 1110012E06Rik; AA589443; AT132 antibody; Atp13a2; ATPase 13A2; ATPase type 13A2; Cation-transporting ATPase 13A2; CLN12; HSA9947; HSA9947 antibody; KRPPD; PARK9; probable cation-transporting ATPase 13A2; putative ATPase; RGD1307977; RP1-37C10.4 |
| Symboles de gène | ATP13A2 |
| Afficher plus de résultats |