missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATP Synthase B1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA527826

Numéro de catalogue. PIPA527826


Il en reste null

Description

Description

Recommended positive controls: Molt-4, mouse brain. Predicted reactivity: Mouse (81%), Rat (80%), Rhesus Monkey (89%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel seems to have nine subunits (a, b, c, d, e, f, g, F6 and 8). This gene encodes the b subunit of the proton channel.
Spécifications

Identifiants chimiques

ATP Synthase B1
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human, Mouse
ATP5PB
ATP synthase B chain, mitochondrial; ATP synthase F(0) complex subunit B1, mitochondrial; ATP synthase peripheral stalk-membrane subunit b; ATP synthase proton-transporting mitochondrial F(0) complex subunit B1; ATP synthase subunit b; ATP synthase subunit b, mitochondrial; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit b, isoform 1; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit B1; ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit B1; Atp5f; ATP5F1; Atp5pb; ATPase subunit b; C76477; cell proliferation-inducing protein 47; H+-ATP synthase subunit b; PIG47
ATP5PB
Primary
Antigen affinity chromatography
0.45 mg/mL
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P24539, Q9CQQ7
11950, 515
Recombinant protein encompassing a sequence within the center region of human ATP synthase B1. The exact sequence is proprietary.
Antibody

Spécifications

ATP Synthase B1
Polyclonal
Unconjugated
ATP5PB
ATP synthase B chain, mitochondrial; ATP synthase F(0) complex subunit B1, mitochondrial; ATP synthase peripheral stalk-membrane subunit b; ATP synthase proton-transporting mitochondrial F(0) complex subunit B1; ATP synthase subunit b; ATP synthase subunit b, mitochondrial; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit b, isoform 1; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit B1; ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit B1; Atp5f; ATP5F1; Atp5pb; ATPase subunit b; C76477; cell proliferation-inducing protein 47; H+-ATP synthase subunit b; PIG47
Rabbit
Antigen affinity chromatography
RUO
11950, 515
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.45 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P24539, Q9CQQ7
ATP5PB
Recombinant protein encompassing a sequence within the center region of human ATP synthase B1. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse
Antibody
IgG
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions