missing translation for 'onlineSavingsMsg'
Learn More

Atorvastatin Calcium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013983
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013983 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013983 Supplier Merck Emd Millipore Supplier No. PHR14221G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Atorvastatin Calcium is a synthetic cholesterol lowering agent, which acts as an inhibitor of 3-hydroxy-3-methyl glutaryl coenzyme A reductase. It is widely used in combination with amlodipine besylate as a medication in the treatment of vasospastic angina, hypertension, chronic stable angina and primary dysbetalipoproteinemia.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 344423-98-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C66H68 CaF2N4O10 · 3H2 O
Synonym Atorvastatin hemicalcium salt sesquihydrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 1209.39
Grade Certified Reference Material