missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATIC Monoclonal Antibody (F38 P7 H9)

Description
MA1-086 detects the ATIC in human, rat, Xenopus, Drosophila, and mouse cells. MA1-086 has been successfully used in Western blot and immunohistochemistry (parrafin) procedures. By Western blot, this antibody detects a ~64.6 kDa protein. The MA1-086 immunogen is a synthetic peptide, conjugated to Ovalbumin, corresponding to residues A(583) H T N L R L F H H(592) of human ATIC..
ATIC is a 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase which helps catalyze the penultimate and final steps of the de novo purine biosynthetic pathway. The human ATIC cDNA encodes a deduced 591-amino acid protein that is 81% identical to the chicken sequence.
Spécifications
Spécifications
| Antigène | ATIC |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | F38 P7 H9 |
| Concentration | 1 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 1 mg/mL BSA and 0.05% sodium azide |
| Gène | ATIC |
| Numéro d’acquisition du gène | O35567, P31939, Q9CWJ9 |
| Alias du gène | 153761_at; 2610509C24Rik; 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase; 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase; 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase L homeolog; 5-aminoimidazole-4-carboxamide-1-beta-D-ribonucleotide transformylase/inosinicase; AA536954; AICAR; AICAR formyltransferase/IMP cyclohydrolase bifunctional enzyme; AICAR transformylase; AICARFT; AICARFT/IMPCHASE; atic; atic.L; AW212393; BEST:GH11240; Bifunctional purine biosynthesis protein PURH; Bifunctional purine biosynthesis protein PURH, N-terminally processed; CG11089; CG11089-PA; CG11089-PB; CT30991; Dmel\CG11089; Dmel_CG11089; epididymis secretory sperm binding protein Li 70p; HEL-S-70p; IMP cyclohydrolase; IMP synthase; IMPCHASE; Inosinicase; OK/SW-cl.86; Phosphoribosylaminoimidazolecarboxamide formyltransferase; phosphoribosylaminoimidazolecarboxamide formyltransferase/IMP cyclohydrolase; PURH; RE29555p; uncharacterized protein; XELAEV_18044849mg |
| Afficher plus de résultats |